APO456791666 Apollo Scientific Ltd

(3S)-3-Amino-1-benzylpiperidine

  • MDL number MFCD03093375
  • Cas Number : 168466-85-1
  • Molecular Weight : 190.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3S)-1-benzyl-3-piperidinamine
Inchi Code 1S/C12H18N2/c13-12-7-4-8-14(10-12)9-11-5-2-1-3-6-11/h1-3,5-6,12H,4,7-10,13H2/t12-/m0/s1
Inchi Key HARWNWOLWMTQCC-LBPRGKRZSA-N

Description

Synonym (3S)-1-Benzylpiperidin-3-amine
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3s-3-amino-1-benzylpiperidine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3s-3-amino-1-benzylpiperidine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart