APO456791681 Apollo Scientific Ltd

4-Cyclohexylphenylglyoxal hydrate

  • MDL number MFCD08272329
  • Cas Number : 99433-89-3
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-cyclohexylphenylglyoxal-hydrate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-cyclohexylphenylglyoxal-hydrate
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart