APO456791734 Apollo Scientific Ltd

4-Nitro-1,5,6,7-tetrahydro-5,5,7,7-tetramethylcyclopenta[f]indazole

  • MDL number MFCD11109510
  • Cas Number : 1209368-38-6
  • Molecular Weight : 259.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-nitro-1567-tetrahydro-5577-tetramethylcyclopentafindazole

Safety Information

signalword Warning
pcodes P260;P280;P312
MSDS https://store.apolloscientific.co.uk/product/4-nitro-1567-tetrahydro-5577-tetramethylcyclopentafindazole
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart