APO456791792 Apollo Scientific Ltd

Hydroxyacetaldehyde diethyl acetal

  • MDL number MFCD00051486
  • Cas Number : 621-63-6
  • Molecular Weight : 134.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 62
Iupac Name 2,2-diethoxyethanol
Inchi Code 1S/C6H14O3/c1-3-8-6(5-7)9-4-2/h6-7H,3-5H2,1-2H3
Inchi Key IKKUKDZKIIIKJK-UHFFFAOYSA-N

Description

Melting Point 62
Synonym 2,2-Diethoxyethan-1-ol, 1,1-Diethoxy-2-hydroxyethane, Glycolaldehyde diethyl acetal
Flash Point 166-167
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/hydroxyacetaldehyde-diethyl-acetal

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/hydroxyacetaldehyde-diethyl-acetal
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart