APO456791812 Apollo Scientific Ltd

2-Bromothiophene-3-carboxaldehyde

  • MDL number MFCD09025880
  • Cas Number : 1860-99-7
  • Molecular Weight : 191.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-3-thiophenecarbaldehyde
Inchi Code 1S/C5H3BrOS/c6-5-4(3-7)1-2-8-5/h1-3H
Inchi Key MMEGVQIGIBCTHI-UHFFFAOYSA-N

Description

Synonym 2-Bromo-3-formylthiophene, 2-Bromo-3-thenaldehyde
Purity 97%
Flash Point 60/0.3mm
Storage temp. Store at 2-8
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromothiophene-3-carboxaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromothiophene-3-carboxaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart