APO456791893 Apollo Scientific Ltd

5-Cyano-3-iodo-1H-indole, N-BOC protected

  • MDL number MFCD05864767
  • Cas Number : 864685-26-7
  • Molecular Weight : 368.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 5-cyano-3-iodo-1H-indole-1-carboxylate
Inchi Code 1S/C14H13IN2O2/c1-14(2,3)19-13(18)17-8-11(15)10-6-9(7-16)4-5-12(10)17/h4-6,8H,1-3H3
Inchi Key GADLXNFRAAUWAD-UHFFFAOYSA-N

Description

Synonym 5-Cyano-3-iodo-1H-indole-1-carboxylic acid tert-butyl ester
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-cyano-3-iodo-1h-indole-n-boc-protected

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-cyano-3-iodo-1h-indole-n-boc-protected
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart