APO456791923 Apollo Scientific Ltd

2,4,6-Trichloropyrimidine-5-carboxaldehyde

  • MDL number MFCD02257700
  • Cas Number : 50270-27-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Formyl-2,4,6-trichloropyrimidine, 5-Formyl-2,4,6-trichloro-1,3-diazine
Storage temp. Store at 2-8
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=46720

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=46720
Availability details for
Quantity
  • Availability
  • Add to Cart