APO456791937 Apollo Scientific Ltd

Ethyl 4-amino-2-chlorobenzoate

  • MDL number MFCD09034640
  • Cas Number : 16017-69-9
  • Molecular Weight : 199.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109-111
Synonym 3-Chloro-4-(ethoxycarbonyl)aniline
Purity 95
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-4-amino-2-chlorobenzoate

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-4-amino-2-chlorobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart