APO456791954 Apollo Scientific Ltd

5-Bromo-2-chloro-6-cyclopropylnicotinonitrile

  • MDL number MFCD14584776
  • Cas Number : 1221792-18-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Bromo-2-chloro-6-cyclopropylpyridine-3-carbonitrile
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-chloro-6-cyclopropylnicotinonitrile

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-chloro-6-cyclopropylnicotinonitrile
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart