APO456792052 Apollo Scientific Ltd

Methyl pyrimidine-4-carboxylate

  • MDL number MFCD00234184
  • Cas Number : 2450-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63-71
Synonym 4-(Methoxycarbonyl)pyrimidine, 4-(Methoxycarbonyl)-1,3-diazine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/methyl-pyrimidine-4-carboxylate

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-pyrimidine-4-carboxylate
pictograms GHS05;GHS07
hstatements H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart