APO456792069 Apollo Scientific Ltd

3-[(Pyrrolidin-1-yl)methyl]benzeneboronic acid, pinacol ester

  • MDL number MFCD08741425
  • Cas Number : 884507-45-3
  • Molecular Weight : 287.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]pyrrolidine
Inchi Code 1S/C17H26BNO2/c1-16(2)17(3,4)21-18(20-16)15-9-7-8-14(12-15)13-19-10-5-6-11-19/h7-9,12H,5-6,10-11,13H2,1-4H3
Inchi Key AKQLICZDOCQKRA-UHFFFAOYSA-N

Description

Synonym 1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]pyrrolidine
Purity 97%
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-pyrrolidin-1-ylmethylbenzeneboronic-acid-pinacol-ester

Safety Information

signalword Danger
pcodes P280;P309+P311
MSDS https://store.apolloscientific.co.uk/product/3-pyrrolidin-1-ylmethylbenzeneboronic-acid-pinacol-ester
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart