APO456792072 Apollo Scientific Ltd

Lithium 2-[(morpholin-4-yl)methyl]benzoate

  • MDL number MFCD08690277
  • Cas Number : 904696-60-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >300(dec.)
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=32799

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=32799
pictograms GHS07
hstatements H302;H312;H332
Availability details for
Quantity
  • Availability
  • Add to Cart