APO456792081 Apollo Scientific Ltd

3-[(2,4-Dimethylphenyl)carbamoyl]benzeneboronic acid

  • MDL number MFCD09027261
  • Cas Number : 957060-98-9
  • Molecular Weight : 269.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-[(2,4-dimethylanilino)carbonyl]phenylboronic acid
Inchi Code 1S/C15H16BNO3/c1-10-6-7-14(11(2)8-10)17-15(18)12-4-3-5-13(9-12)16(19)20/h3-9,19-20H,1-2H3,(H,17,18)
Inchi Key ZVBRLWSDAGYMQQ-UHFFFAOYSA-N

Description

Synonym 3-Borono-N-(2,4-dimethylphenyl)benzamide
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-24-dimethylphenylcarbamoylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/3-24-dimethylphenylcarbamoylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart