APO456792095 Apollo Scientific Ltd

4-Methoxybenzene-1,2-diamine

  • MDL number MFCD00047837
  • Cas Number : 102-51-2
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 46-50
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-methoxybenzene-12-diamine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-methoxybenzene-12-diamine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart