APO456792107 Apollo Scientific Ltd

4-Amino-3-nitrobenzenesulphonamide

  • MDL number MFCD00116612
  • Cas Number : 2360-19-2
  • Molecular Weight : 217.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 204-210
Iupac Name 4-amino-3-nitrobenzenesulfonamide
Inchi Code 1S/C6H7N3O4S/c7-5-2-1-4(14(8,12)13)3-6(5)9(10)11/h1-3H,7H2,(H2,8,12,13)
Inchi Key DZJCUJCWFQPICK-UHFFFAOYSA-N

Description

Synonym 2-Nitro-4-sulphamoylaniline, 4-(Aminosulphonyl)-2-nitroaniline
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 204-210
Product Link https://store.apolloscientific.co.uk/product/4-amino-3-nitrobenzenesulphonamide

Safety Information

signalword Warning
pcodes P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-3-nitrobenzenesulphonamide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart