APO456792258 Apollo Scientific Ltd

2-(Bromoacetyl)-1,3-benzothiazole

  • MDL number MFCD03659698
  • Cas Number : 54223-20-0
  • Molecular Weight : 256.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86-87
Synonym 1-(1,3-Benzothiazol-2-yl)-2-bromoethan-1-one, 2-(2-Bromoethanoyl)-1,3-benzothiazole
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromoacetyl-13-benzothiazole

Safety Information

signalword Danger
pcodes P260;P280;P310
MSDS https://store.apolloscientific.co.uk/product/2-bromoacetyl-13-benzothiazole
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart