APO456792262 Apollo Scientific Ltd

Benzo[b]furan-5-carboxylic acid

  • MDL number MFCD01006742
  • Cas Number : 90721-27-0
  • Molecular Weight : 162.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 189-190
Iupac Name 1-benzofuran-5-carboxylic acid
Inchi Code 1S/C9H6O3/c10-9(11)7-1-2-8-6(5-7)3-4-12-8/h1-5H,(H,10,11)
Inchi Key GTWXSZIQNTUNKR-UHFFFAOYSA-N

Description

Synonym 1-Benzofuran-5-carboxylic acid, 5-Carboxybenzo[b]furan
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 189-190
Product Link https://store.apolloscientific.co.uk/product/benzobfuran-5-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/benzobfuran-5-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart