APO456792271 Apollo Scientific Ltd

2,1,3-Benzothiadiazole-5-carboxaldehyde

  • MDL number MFCD00457949
  • Cas Number : 71605-72-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89-95
Synonym 5-Formyl-2,1,3-benzothiadiazole
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=19333

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=19333
pictograms GHS07
hstatements H302;H312;H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart