APO456792272 Apollo Scientific Ltd

4-(Bromomethyl)-5-phenyl-1,3-oxazole

  • MDL number MFCD03407316
  • Cas Number : 368869-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50-52
Synonym [4-(Bromomethyl)-1,3-oxazol-5-yl]benzene
Purity 95
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=19345

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=19345
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart