APO456792416 Apollo Scientific Ltd

Ethyl 2-amino-4-propylpyrimidicne-5-carboxylate

  • MDL number MFCD00052079
  • Cas Number : 127957-83-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=20542

Safety Information

signalword Warning
pcodes P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=20542
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart