APO456792418 Apollo Scientific Ltd

Methyl 4-aminothiophene-3-carboxylate

  • MDL number MFCD00130102
  • Cas Number : 69363-85-5
  • Molecular Weight : 157.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 35-38
Synonym 3-Amino-4-(methoxycarbonyl)thiophene, Methyl 4-amino-3-thenoate
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-4-aminothiophene-3-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-4-aminothiophene-3-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart