APO456792425 Apollo Scientific Ltd

1,4-Diiodo-2,5-dimethoxybenzene

  • MDL number MFCD00461369
  • Cas Number : 51560-21-5
  • Molecular Weight : 389.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 171-173
Iupac Name 1,4-diiodo-2,5-dimethoxybenzene
Inchi Code 1S/C8H8I2O2/c1-11-7-3-6(10)8(12-2)4-5(7)9/h3-4H,1-2H3
Inchi Key GLVOXVCTGAISRY-UHFFFAOYSA-N

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 171-173
Product Link https://store.apolloscientific.co.uk/product/14-diiodo-25-dimethoxybenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/14-diiodo-25-dimethoxybenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart