APO456792452 Apollo Scientific Ltd

4-Methoxythiophene-3-carboxylic acid

  • MDL number MFCD00067997
  • Cas Number : 71050-40-3
  • Molecular Weight : 158.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 101-105
Iupac Name 4-methoxy-3-thiophenecarboxylic acid
Inchi Code 1S/C6H6O3S/c1-9-5-3-10-2-4(5)6(7)8/h2-3H,1H3,(H,7,8)
Inchi Key KEYPLCJVNVJJQK-UHFFFAOYSA-N

Description

Synonym 3-Carboxy-4-methoxythiophene
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 101-105
Product Link https://store.apolloscientific.co.uk/product/4-methoxythiophene-3-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-methoxythiophene-3-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart