APO456792487 Apollo Scientific Ltd

6,6-Dimethyl-3-(methylthio)-4-oxo-4,5,6,7-tetrahydrobenzo[c]thiophene-1-carbonitrile

  • MDL number MFCD00084980
  • Cas Number : 175202-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169-171
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=21478

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=21478
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart