APO456792552 Apollo Scientific Ltd

3-Hydroxythiophene-2-carboxylic acid

  • MDL number MFCD09414719
  • Cas Number : 5118-07-0
  • Molecular Weight : 144.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 106-108
Iupac Name 3-hydroxy-2-thiophenecarboxylic acid
Inchi Code 1S/C5H4O3S/c6-3-1-2-9-4(3)5(7)8/h1-2,6H,(H,7,8)
Inchi Key RVDUJCGAAPQDBO-UHFFFAOYSA-N

Description

Synonym 2-Carboxy-3-hydroxythiophene, 3-Hydroxy-2-thenoic acid
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 106-108
Product Link https://store.apolloscientific.co.uk/product/3-hydroxythiophene-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-hydroxythiophene-2-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart