APO456792659 Apollo Scientific Ltd

2-(3,5-Dichlorophenyl)propene

  • MDL number MFCD00035987
  • Cas Number : 68575-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=36903

Safety Information

signalword Warning
pcodes P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=36903
pictograms GHS07
hstatements H302+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart