APO456792672 Apollo Scientific Ltd

3,5-Dichloro-4-hydroxybenzaldehyde

  • MDL number MFCD00017605
  • Cas Number : 2314-36-5
  • Molecular Weight : 191.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 156-158
Iupac Name 3,5-dichloro-4-hydroxybenzaldehyde
Inchi Code 1S/C7H4Cl2O2/c8-5-1-4(3-10)2-6(9)7(5)11/h1-3,11H
Inchi Key LIYGCLJYTHRBQV-UHFFFAOYSA-N

Description

Synonym 2,6-Dichloro-4-formylphenol
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 156-158
Product Link https://store.apolloscientific.co.uk/product/35-dichloro-4-hydroxybenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/35-dichloro-4-hydroxybenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart