APO456792674 Apollo Scientific Ltd

Methyl 5-bromo-2-ethoxynicotinate

  • MDL number MFCD09842613
  • Cas Number : 1031927-15-7
  • Molecular Weight : 260.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 5-bromo-2-ethoxynicotinate
Inchi Code 1S/C9H10BrNO3/c1-3-14-8-7(9(12)13-2)4-6(10)5-11-8/h4-5H,3H2,1-2H3
Inchi Key DRYKICBAPXKRKF-UHFFFAOYSA-N

Description

Synonym Methyl 5-bromo-2-ethoxypyridine-3-carboxylate, 5-Bromo-2-ethoxy-3-(methoxycarbonyl)pyridine
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-5-bromo-2-ethoxynicotinate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-5-bromo-2-ethoxynicotinate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart