APO456792675 Apollo Scientific Ltd

6-Bromo-8-nitroimidazo[1,2-a]pyridine

  • MDL number MFCD09836167
  • Cas Number : 1031927-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=42619

Safety Information

signalword Warning
pcodes P280;P301+P312;P305+P351+P338
MSDS http://www.apolloscientific.co.uk/display_item.php?id=42619
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart