APO456792692 Apollo Scientific Ltd

4-[(N,N-Dimethylamino)methyl]benzeneboronic acid pinacol ester hydrochloride

  • MDL number MFCD07771993
  • Cas Number : 1073371-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-nn-dimethylaminomethylbenzeneboronic-acid-pinacol-ester-hydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-nn-dimethylaminomethylbenzeneboronic-acid-pinacol-ester-hydrochloride
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart