APO456792794 Apollo Scientific Ltd

2-Chloro-6-methylphenyl isothiocyanate

  • MDL number MFCD00039643
  • Cas Number : 19241-34-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Chloro-2-isothiocyanatotoluene, 1-Chloro-2-isothiocyanato-3-methylbenzene
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 134-135/10mm
Product Link https://store.apolloscientific.co.uk/product/2-chloro-6-methylphenyl-isothiocyanate

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-6-methylphenyl-isothiocyanate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart