APO456792904 Apollo Scientific Ltd

3-Chloro-6-hydroxyisoquinoline

  • MDL number MFCD22380493
  • Cas Number : 1374652-53-5
  • Molecular Weight : 179.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-chloro-6-isoquinolinol
Inchi Code 1S/C9H6ClNO/c10-9-4-7-3-8(12)2-1-6(7)5-11-9/h1-5,12H
Inchi Key UGPQRQFIFJTENM-UHFFFAOYSA-N

Description

Synonym 3-Chloroisoquinolin-6-ol, 3-Chloro-6-hydroxy-2-azanaphthalene
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloro-6-hydroxyisoquinoline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-6-hydroxyisoquinoline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart