APO456792930 Apollo Scientific Ltd

2-Bromo-4-methoxybenzaldehyde

  • MDL number MFCD09788636
  • Cas Number : 43192-31-0
  • Molecular Weight : 215.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-4-methoxybenzaldehyde
Inchi Code 1S/C8H7BrO2/c1-11-7-3-2-6(5-10)8(9)4-7/h2-5H,1H3
Inchi Key ODISAUHBLBVQKC-UHFFFAOYSA-N

Description

Synonym 3-Bromo-4-formylanisole, 2-Bromo-p-anisaldehyde
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-4-methoxybenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-4-methoxybenzaldehyde
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart