APO456792945 Apollo Scientific Ltd

6-Hydroxy-2-azaspiro[3.3]heptane, N-BOC protected

  • MDL number MFCD15071431
  • Cas Number : 1147557-97-8
  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl 6-hydroxy-2-azaspiro[3.3]heptane-2-carboxylate, 2-(tert-Butoxycarbonyl)-6-hydroxy-2-azaspiro[3.3]heptane
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-hydroxy-2-azaspiro33heptane-n-boc-protected

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-hydroxy-2-azaspiro33heptane-n-boc-protected
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart