APO456792963 Apollo Scientific Ltd

2-Methylbutyl isothiocyanate

  • MDL number MFCD00060414
  • Cas Number : 4404-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Isothiocyanato-2-methylbutane
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 84-86/15mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=26370

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=26370
pictograms GHS05;GHS06
hstatements H301+H311+H331;H314
Availability details for
Quantity
  • Availability
  • Add to Cart