APO456792972 Apollo Scientific Ltd

4'-Bromo-2-hydroxyacetophenone

  • MDL number MFCD00174275
  • Cas Number : 3343-45-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137-142
Synonym 1-(4-Bromophenyl)-2-hydroxyethan-1-one
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2-hydroxyacetophenone

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2-hydroxyacetophenone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart