APO456792998 Apollo Scientific Ltd

16-(tert-Butoxy)-16-oxohexadecanoic acid

  • MDL number MFCD23136039
  • Cas Number : 843666-27-3
  • Molecular Weight : 342.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 16-tert-butoxy-16-oxohexadecanoic acid
Inchi Code 1S/C20H38O4/c1-20(2,3)24-19(23)17-15-13-11-9-7-5-4-6-8-10-12-14-16-18(21)22/h4-17H2,1-3H3,(H,21,22)
Inchi Key HXJICNOLPKEOLU-UHFFFAOYSA-N

Description

Synonym 16-(tert-Butoxy)-16-oxopalmitic acid, tert-Butyl 15-carboxypentadecanoate
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/16-tert-butoxy-16-oxohexadecanoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/16-tert-butoxy-16-oxohexadecanoic-acid
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart