APO456793000 Apollo Scientific Ltd

18-(tert-Butoxy)-18-oxooctadecanoic acid

  • MDL number MFCD09991735
  • Cas Number : 843666-40-0
  • Molecular Weight : 370.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 18-tert-butoxy-18-oxooctadecanoic acid
Inchi Code 1S/C22H42O4/c1-22(2,3)26-21(25)19-17-15-13-11-9-7-5-4-6-8-10-12-14-16-18-20(23)24/h4-19H2,1-3H3,(H,23,24)
Inchi Key WDUQJXKBWRNMKI-UHFFFAOYSA-N

Description

Synonym 18-(tert-Butoxy)-18-oxostearic acid, tert-Butyl 17-carboxyheptadecanoate
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/18-tert-butoxy-18-oxooctadecanoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/18-tert-butoxy-18-oxooctadecanoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart