APO456793081 Apollo Scientific Ltd

5-(Aminomethyl)-1,3-thiazole

  • MDL number MFCD02853613
  • Cas Number : 161805-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (1,3-Thiazol-5-yl)methylamine
Purity 95
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=44867

Safety Information

signalword Danger
pcodes P271;P280;P260
MSDS http://www.apolloscientific.co.uk/display_item.php?id=44867
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart