APO456793101 Apollo Scientific Ltd

Methyl 1H-pyrazole-4-carboxylate

  • MDL number MFCD00297363
  • Cas Number : 51105-90-9
  • Molecular Weight : 126.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 1H-pyrazole-4-carboxylate
Inchi Code 1S/C5H6N2O2/c1-9-5(8)4-2-6-7-3-4/h2-3H,1H3,(H,6,7)
Inchi Key VFTZKSMAJVLWOV-UHFFFAOYSA-N

Description

Synonym 4-(Methoxycarbonyl)-1H-pyrazole
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-1h-pyrazole-4-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-1h-pyrazole-4-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart