APO456793108 Apollo Scientific Ltd

Azetidin-3-one, N-CBZ protected

  • MDL number MFCD09954923
  • Cas Number : 105258-93-3
  • Molecular Weight : 205.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Benzyl 3-oxoazetidine-1-carboxylate, 1-[(Benzyloxy)carbonyl]-3-oxoazetidine
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/azetidin-3-one-n-cbz-protected

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/azetidin-3-one-n-cbz-protected
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart