APO456793168 Apollo Scientific Ltd

4-(tert-Butoxy)benzeneboronic acid, pinacol ester

  • MDL number MFCD08741438
  • Cas Number : 938063-51-5
  • Molecular Weight : 276.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl ether
Inchi Code 1S/C16H25BO3/c1-14(2,3)18-13-10-8-12(9-11-13)17-19-15(4,5)16(6,7)20-17/h8-11H,1-7H3
Inchi Key MJXCVJJOCYCOQE-UHFFFAOYSA-N

Description

Synonym 2-(4-tert-Butoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-tert-butoxybenzeneboronic-acid-pinacol-ester

Safety Information

signalword Warning
pcodes P260;P280;P312
MSDS https://store.apolloscientific.co.uk/product/4-tert-butoxybenzeneboronic-acid-pinacol-ester
pictograms GHS07
hstatements H302+H312+H332;H319
Availability details for
Quantity
  • Availability
  • Add to Cart