APO456793252 Apollo Scientific Ltd

5-Hydroxy-2,1,3-benzoxadiazole

  • MDL number MFCD00173849
  • Cas Number : 768-09-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104-107
Synonym 2,1,3-Benzoxadiazol-5-ol, 5-Hydroxybenzofurazan
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=14811

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=14811
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart