APO456793276 Apollo Scientific Ltd

4-[(Ethylcarbamoyl)amino]benzeneboronic acid, pinacol ester

  • MDL number MFCD08689523
  • Cas Number : 874291-00-6
  • Molecular Weight : 290.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 194-196
Iupac Name N-ethyl-N'-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]urea
Inchi Code 1S/C15H23BN2O3/c1-6-17-13(19)18-12-9-7-11(8-10-12)16-20-14(2,3)15(4,5)21-16/h7-10H,6H2,1-5H3,(H2,17,18,19)
Inchi Key BYHDTZHNPIWVOP-UHFFFAOYSA-N

Description

Synonym 4-(3-Ethylureido)benzeneboronic acid, pinacol ester
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 194-196
Product Link https://store.apolloscientific.co.uk/product/4-ethylcarbamoylaminobenzeneboronic-acid-pinacol-ester

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-ethylcarbamoylaminobenzeneboronic-acid-pinacol-ester
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart