APO456793329 Apollo Scientific Ltd

1,3-Dihydro-1-phenyl-2H-indol-2-one

  • MDL number MFCD00234850
  • Cas Number : 3335-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123-125
Synonym 1-Phenylindolin-2-one, 1-Phenyl-2-oxindole
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/13-dihydro-1-phenyl-2h-indol-2-one

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/13-dihydro-1-phenyl-2h-indol-2-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart