APO456793355 Apollo Scientific Ltd

2,4,6-Trimethoxyaniline

  • MDL number MFCD00017165
  • Cas Number : 14227-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=38815

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=38815
Availability details for
Quantity
  • Availability
  • Add to Cart