APO456793395 Apollo Scientific Ltd

1-(2-Bromobenzyl)piperidine

  • MDL number MFCD07775745
  • Cas Number : 1459-22-9
  • Molecular Weight : 254.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-bromobenzyl)piperidine
Inchi Code 1S/C12H16BrN/c13-12-7-3-2-6-11(12)10-14-8-4-1-5-9-14/h2-3,6-7H,1,4-5,8-10H2
Inchi Key YHPWAILJEPQMRO-UHFFFAOYSA-N

Description

Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-2-bromobenzylpiperidine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-2-bromobenzylpiperidine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart