APO456793440 Apollo Scientific Ltd

5-Amino-3-cyclopropyl-1H-pyrazole

  • MDL number MFCD00067985
  • Cas Number : 175137-46-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Cyclopropyl-1H-pyrazol-5-amine, (5-Amino-1H-pyrazol-3-yl)cyclopropane
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=31538

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=31538
pictograms GHS07
hstatements H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart