APO456793489 Apollo Scientific Ltd

7-Hydroxybenzo[b]furan

  • MDL number MFCD03788248
  • Cas Number : 4790-81-2
  • Molecular Weight : 134.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 42-43
Iupac Name 1-benzofuran-7-ol
Inchi Code 1S/C8H6O2/c9-7-3-1-2-6-4-5-10-8(6)7/h1-5,9H
Inchi Key WXVRAHOQLYAQCR-UHFFFAOYSA-N

Description

Synonym 1-Benzofuran-7-ol, Benzo[b]furan-7-ol
Purity 95%
Flash Point 240
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 42-43
Product Link https://store.apolloscientific.co.uk/product/7-hydroxybenzobfuran

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/7-hydroxybenzobfuran
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart