APO456793490 Apollo Scientific Ltd

2,4,6-Trimethoxybenzeneboronic acid

  • MDL number MFCD01114642
  • Cas Number : 135159-25-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96-100
Purity 97
Storage temp. Store at -20
Physical Form Solid
Shipping Temperature Shipped with Ice Packs
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/246-trimethoxybenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/246-trimethoxybenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart